PharmaDispo Disposable Powder Samplers
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5393-8000 5393-8001 5393-8010 5393-8011 5393-8020 5393-8021 5393-8030 5393-8031 5393-8040 5393-8041
BURK5393-8000EA 0 EUR
BURK5393-8000 BURK5393-8001 BURK5393-8010 BURK5393-8011 BURK5393-8020 BURK5393-8021 BURK5393-8030 BURK5393-8031 BURK5393-8040 BURK5393-8041
PharmaDispo Disposable Powder Samplers
Monsternemers Monsternemers voor eenmalig gebruik
PharmaDispo disposable powder sampler is a multi-target sampler that takes three separate target point samples with a single sampling.
  • Take small individual samples from different layers
  • Sterile versions are additionally gamma sterilised
  • Easy to use: Sliding mechanism with end stop for user-friendly operation
  • Hygienic and safe: Clean room manufactured, individually packaged, specifically designed for single use
The three separate sampling chambers of the PharmaDispo allow simultaneous sampling from different layers or zones. This allows the materialMore Product Information