Drum funnels
Leverancier: BURKLE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9602-0350 9602-0250 9602-0400 9602-0450 9602-0200
BURK9602-0350EA 34.9 EUR
BURK9602-0350 BURK9602-0250 BURK9602-0400 BURK9602-0450 BURK9602-0200
Drum funnels
Trechters Filtertrechters Trechters voor vaten
Made of PP, translucent.
  • Very good chemical resistance
  • Very good temperature resistance
  • Quick and easy to clean
For filling containers with large quantities of liquid or powdered substances quickly.
Order Now
Specificaties
Environmentally Preferable