Anti-p-4E-BP1 Rabbit Polyclonal Antibody [clone: T69]

Leverancier: Bioworld Technology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS4003
BWRLBS4003EA 284 EUR
BWRLBS4003
Anti-p-4E-BP1 Rabbit Polyclonal Antibody [clone: T69]
Antilichamen
Synthetic phosphopeptide derived from human 4E-BP1 around the phosphorylation site of Threonine 69.

p-4E-BP1 (T69) pAb detects endogenous levels of 4E-BP1 protein only when phosphorylated at Thr69.

Type: Primary
Antigen: p-4E-BP1
Clonality: Polyclonal
Clone: T69
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR