Anti-p-WNK1 Rabbit Polyclonal Antibody [clone: T60]

Leverancier: Bioworld Technology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS4208
BWRLBS4208EA 284 EUR
BWRLBS4208
Anti-p-WNK1 Rabbit Polyclonal Antibody [clone: T60]
Antilichamen
Synthetic phosphopeptide derived from human WNK1 around the phosphorylation site of Threonine 60.

p-WNK1 (T60) pAb detects endogenous levels of WNK1 protein only when phosphorylated at Thr60.

Type: Primary
Antigen: p-WNK1
Clonality: Polyclonal
Clone: T60
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR