Anti-p-p70 S6K Rabbit Polyclonal Antibody [clone: T389]

Leverancier: Bioworld Technology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS4440
BWRLBS4440EA 284 EUR
BWRLBS4440
Anti-p-p70 S6K Rabbit Polyclonal Antibody [clone: T389]
Antilichamen
Synthetic phosphopeptide derived from human p70 S6K around the phosphorylation site of Threonine 389.

p-p70 S6K (T389) pAb detects endogenous levels of p70 S6 kinase only when phosphorylated at Thr389. This antibody also detects p85 S6 kinase when phosphorylated at Thr412.

Type: Primary
Antigen: p-p70 S6K
Clonality: Polyclonal
Clone: T389
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR