Anti-CASP10 Rabbit Polyclonal Antibody

Leverancier: Bioworld Technology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS5642
BWRLBS5642EA 262 EUR
BWRLBS5642
Anti-CASP10 Rabbit Polyclonal Antibody
Antilichamen
Synthetic peptide, corresponding to amino acids 206-251 of Human Caspase 10.

Caspase 10 pAb detects endogenous levels of Caspase 10 protein.

Type: Primary
Antigen: CASP10
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR