Anti-ENPP3 Rabbit Polyclonal Antibody

Leverancier: Bioworld Technology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS60438
BWRLBS60438EA 262 EUR
BWRLBS60438
Anti-ENPP3 Rabbit Polyclonal Antibody
Antilichamen
A synthetic peptide corresponding to residues in Human ENPP3.

ENPP3 pAb detects endogenous levels of ENPP3 protein.

Type: Primary
Antigen: ENPP3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Specificaties

antigeen symbool ENPP3
Environmentally Preferable
Type antilichaam Primair
Clonality Polyklonaal
Reactivity Human,Rat,Mouse
Host Rabbit
Isotype
Western blot Yes

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR