Anti-Dok-1 Rabbit Polyclonal Antibody

Leverancier: Bioworld Technology
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS7103
BWRLBS7103EA 341 EUR
BWRLBS7103
Anti-Dok-1 Rabbit Polyclonal Antibody
Antilichamen
Recombinant full length Human Dok-1.

Dok-1 pAb detects endogenous levels of Dok-1 protein.

Type: Primary
Antigen: Dok-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR