Multi-gas detector, GasAlert Max XT II

Leverancier: BW TECHNOLOGIES
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

XT-0W0M-Y-EU XT-X000-Y-EU XT-XW00-Y-EU XT-XW0M-Y-EU XT-XWHM-Y-EU XT-0W00-Y-EU XT-X00M-Y-EU XT-0W00-Y-UK XT-0W0M-Y-UK XT-X000-Y-UK XT-X00M-Y-UK XT-XW00-Y-UK XT-XW0M-Y-UK XT-XWHM-Y-UK
BWTEXT-0W0M-Y-EUEA 1120 EUR
BWTEXT-0W0M-Y-EU BWTEXT-X000-Y-EU BWTEXT-XW00-Y-EU BWTEXT-XW0M-Y-EU 151-0239 BWTEXT-0W00-Y-EU BWTEXT-X00M-Y-EU BWTEXT-0W00-Y-UK BWTEXT-0W0M-Y-UK BWTEXT-X000-Y-UK BWTEXT-X00M-Y-UK BWTEXT-XW00-Y-UK BWTEXT-XW0M-Y-UK BWTEXT-XWHM-Y-UK
Multi-gas detector, GasAlert Max XT II
Gasdetectiesystemen
The rugged GasAlert Max XT II reliably monitors up to four gas hazards and combines straightforward one-button field operation with an integrated sampling pump.

  • Compatible with BW MicroDock II automatic test and calibration station
  • Powered by a rechargeable battery pack
  • GasAlert Max XT II comes with standard event logging and datalogging (accessory connectivity kit required)

Tamper-proof, user-adjustable options enable the instrument to be customised to suit your application.

Leveringsinformatie: Supplied with detector complete with sensors,More Product Information
Order Now

Specificaties

Model GasAlert Max XT II
Environmentally Preferable
Alarm Visual, vibrating, audible (95 dB)
Gebruiksduur accu's 10 hours at −20 °C, 13 hours at 0 °C
b×d×h 7.0×5.2×13.1 cm
Gewicht 316 g

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR