Pestles for 0,5 and 1,5 ml microtubes, Dstroy®-Sticks
Leverancier: BIOZYM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
710388 710389 710398 710399
BZYM710388EA 0 EUR
BZYM710388 BZYM710389 BZYM710398 BZYM710399
Pestles for 0,5 and 1,5 ml microtubes, Dstroy®-Sticks
Vijzels en stampers Stampers
These Dstroy®-Sticks and associated reaction tubes are made of polypropylene.
  • Free from RNases, DNases, DNA and pyrogens
Mostly used in the applications homogenising small amounts of cell materials in a reaction vessel and resuspending protein or DNA precipitates in appropriate buffer solutions.