Strips of 8 PCR tubes 0,2 mland domed cap strips, SoftStrips®
Leverancier: BIOZYM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
710950 710951 710952 710953 710954 710955 710960 710961 710962 710963 710964 710965 710966 711030 711031 711032 711033 711034 711036 711038 711040 711041 711042 711043 711044 711046 711048 711050
BZYM710950EA 0 EUR
BZYM710950 BZYM710951 BZYM710952 BZYM710953 BZYM710954 BZYM710955 BZYM710960 BZYM710961 BZYM710962 BZYM710963 BZYM710964 BZYM710965 BZYM710966 BZYM711030 BZYM711031 BZYM711032 BZYM711033 BZYM711034 BZYM711036 BZYM711038 BZYM711040 BZYM711041 BZYM711042 BZYM711043 BZYM711044 BZYM711046 BZYM711048 BZYM711050
Strips of 8 PCR tubes 0,2 mland domed cap strips, SoftStrips®
Buisjes PCR-buisjes
Tubes without caps. Cap strips must be ordered separately.
  • DNase-, RNase-free
  • DNA-, DNase-, RNase-free (High profile tubes)