Strips of 12 PCR 0,2 ml tubes or domed PCR cap strips, SoftStrips®
Leverancier: BIOZYM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
711060 711061 711062 711063 711064 711068 711070 711071 711073 711074 711076 711078
BZYM711060EA 0 EUR
BZYM711060 BZYM711061 BZYM711062 BZYM711063 BZYM711064 BZYM711068 BZYM711070 BZYM711071 BZYM711073 BZYM711074 BZYM711076 BZYM711078
Strips of 12 PCR 0,2 ml tubes or domed PCR cap strips, SoftStrips®
Buisjes PCR-buisjes
Tubes without caps. Cap strips must be ordered separately.
  • High Profile
  • DNA-, DNase-, RNase-free