PCR tubes, 0,2/0,5 ml, with flat or domed cap, SoftTubes®
Leverancier: BIOZYM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
710900 710901 710902 710903 710904 710906 710908 710910 710911 710912 710913 710914 710915 710916 710918 710921 710922 710923 710924 710926 711000 711001 711002 711003 711004 711006 711008 711080 711081 711082 711083 711084 711086 711088 711090 711091 711092 711093 711094 711096 711098 711610 711615 711650 711655
BZYM710900EA 0 EUR
BZYM710900 BZYM710901 BZYM710902 BZYM710903 BZYM710904 BZYM710906 BZYM710908 BZYM710910 BZYM710911 BZYM710912 BZYM710913 BZYM710914 BZYM710915 BZYM710916 BZYM710918 BZYM710921 BZYM710922 BZYM710923 BZYM710924 BZYM710926 BZYM711000 BZYM711001 BZYM711002 BZYM711003 BZYM711004 BZYM711006 BZYM711008 BZYM711080 BZYM711081 BZYM711082 BZYM711083 BZYM711084 BZYM711086 BZYM711088 BZYM711090 BZYM711091 BZYM711092 BZYM711093 BZYM711094 BZYM711096 BZYM711098 BZYM711610 BZYM711615 BZYM711650 BZYM711655
PCR tubes, 0,2/0,5 ml, with flat or domed cap, SoftTubes®
Buisjes PCR-buisjes
PP, translucent.
  • Excellent temperature resistance
  • Excellent chemical resistance
  • Free from DNase and RNase
With domed or flat cap.