BrightVision+ Anti-IgG (Poly-AP)
Leverancier: IMMUNOLOGIC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
C-DPVB110AP C-DPVB500AP C-DPVB55AP C-DPVB999AP
C-DPVB110APEA 742 EUR
C-DPVB110AP C-DPVB500AP C-DPVB55AP C-DPVB999AP
BrightVision+ Anti-IgG (Poly-AP)
Antilichamen
BrightVision+ histostaining reagents utilise a novel controlled polymerisation technology to prepare polymeric APlinker antibody conjugates. Comparing to conventional biotin-streptavidin based detection kits, BrightVision+ histostaining kits have the advantages of higher amplification power, biotin free and more consistent immunostaining outcomes on archival tissues and on difficult-to-work antibodies.
  • High quality secondary antibody for IHC applications, AP labeled
  • Antigen Specificity: Anti-Mouse IgG (H+L), Anti-Rabbit IgG (H+L), Anti Rat IgG (H+L)
  • Kit
More Product Information