Boorzuur 99,5, granular NF, Macron Fine Chemicals™

Leverancier: Avantor Performance Materials
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid

M.O.C. Category R

C1394-26EA 1985.83 EUR
C1394-26 C1394-20
Boorzuur 99,5, granular NF, Macron Fine Chemicals™
Boorzuur
Formule: B(OH)₃
MW: 61.83 g/mol
Kookpunt: 185
Smeltpunt: 160
Dichtheid: 1.44 g/cm³ (25 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011337
CAS nummer: 10043-35-3
EINECS: 233-139-2
Bereik: 01-2119486683-25
Merck Index: 13,01326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=098dd700154daaf6338ebb270902bb2bc704f27d&vid=6eec7d529090dd4c4b966ed8cd8d057153c5a47b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

GMP Manufactured Product
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical 99.5 - 100.5 %
NF - Assay (H₃BO₃) 99.5 - 100.5 %
NF - Completeness of Solution Passes Test
NF - Identification Passes Test
NF - Loss on Drying ≤0.5 %
NF - Solubility in Alcohol Passes Test
Color of Solution Passes Test
No Class 1,2,3 or other solvents are used or produced in the
manufacturing or purification of the product.
Not for Internal Use
Meets Reagent Specifications for testing USP/NF monographs
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR