γ-Linolenic acid methyl ester

Leverancier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

10006579-100 10006579-50 10006579-250 10006579-1
CAYM10006579-100EA 92.5 EUR
CAYM10006579-100 CAYM10006579-50 CAYM10006579-250 CAYM10006579-1
γ-Linolenic acid methyl ester
γ-Linolenic acid methyl ester
Formule: C₁₉H₃₂O₂
MDL Nummer: MFCD00010455
CAS nummer: 16326-32-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR