Apigenin

Leverancier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

10010275-100 10010275-25 10010275-50 10010275-500
CAYM10010275-100EA 321 EUR
CAYM10010275-100 CAYM10010275-25 CAYM10010275-50 CAYM10010275-500
Apigenin
Apigenin
Formule: C₁₅H₁₀O₅
MDL Nummer: MFCD00006831
CAS nummer: 520-36-5
EINECS: 208-292-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR