Anti-Phosphothreonine Mouse Monoclonal Antibody [clone: 13F9]

Leverancier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

10222-100
CAYM10222-100EA 780 EUR
CAYM10222-100
Anti-Phosphothreonine Mouse Monoclonal Antibody [clone: 13F9]
Antilichamen
Type: Primary
Antigen: Phosphothreonine
Clonality: Monoclonal
Clone: 13F9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR