4-Aminobenzamidoxime

Leverancier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

11053-500 11053-5 11053-1
CAYM11053-500EA 50.3 EUR
CAYM11053-500 CAYM11053-5 CAYM11053-1
4-Aminobenzamidoxime
4-Aminobenzamidoxime
Formule: C₇H₉N₃O
MDL Nummer: MFCD08061151
CAS nummer: 277319-62-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR