2-Amino-4-(4-pyridyl)thiazole

Leverancier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

11058-500 11058-1 11058-5
CAYM11058-500EA 101 EUR
CAYM11058-500 CAYM11058-1 CAYM11058-5
2-Amino-4-(4-pyridyl)thiazole
2-Amino-4-(4-pyridyl)thiazole
Formule: C₈H₇N₃S
MDL Nummer: MFCD00490009
CAS nummer: 30235-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR