Anti-I kappaB-alpha Mouse Monoclonal Antibody [clone: 39A1413]

Leverancier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

13923-1
CAYM13923-1EA 707 EUR
CAYM13923-1
Anti-I kappaB-alpha Mouse Monoclonal Antibody [clone: 39A1413]
Antilichamen
Type: Primary
Antigen: I kappaB-alpha
Clonality: Monoclonal
Clone: 39A1413
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Bovine, Mouse, Pig, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR