3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] ≥98%, crystalline solid

Leverancier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

23124-1 23124-500
CAYM23124-1EA 324 EUR
CAYM23124-1 CAYM23124-500
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] ≥98%, crystalline solid
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide]

3,3'-Dihexyloxacarbocyanine (DiOC₆(3)) is a lipophilic fluorescent dye with excitation/emission spectra of 484/501 nm, respectively.


At high concentrations, DiOC₆(3) accumulates in the endoplasmic reticulum (ER) and has been used to visualise ER in moss, yeast, and muscle cells. At low concentrations, DiOC₆(3) accumulates in mitochondria and has been used to assess mitochondrial dislocations, fusion, and fission in living cells, as well as to visualize cellular apoptosis.


DiOC₆(3) can also be used to assess cell membrane potential, as depolarisation directly correlates with increased fluorescence.2,3 DiOC₆(3) photoexcitation inactivates intracellular organelle movement and inhibits microtubule polymerisation in-vitro.

Formule: C₂₉H₃₇IN₂O₂
Bewaar temperatuur: Vriezer
MDL Nummer: MFCD00011954
CAS nummer: 53213-82-4

Order Now

Specification Test Results

Purity ≥98%
Emission 501 nm
Excitation 484 nm
λmax 486 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR