Tris(3-hydroxypropyltriazolylmethyl)amine ≥95%

Leverancier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: THPTA, Tris[[1-(3-hydroxypropyl)-1H-1,2,3-triazol-4-yl]methyl]amine, 3,3′,3′′-(4,4′,4′′-(Nitrilotris(methylene))tris(1H-1,2,3-triazole-4,1-diyl))tris(propan-1-ol)

Stabilisatie: ≥4 years

35670-100
CAYM35670-100EA 90.3 EUR
CAYM35670-100
Tris(3-hydroxypropyltriazolylmethyl)amine ≥95%
Tris(3-hydroxypropyltriazolylmethyl)amine

A ligand for copper(I)-catalysed azide/alkyne cycloaddition.

Formule: C₁₈H₃₀N₁₀O₃
MW: 434.51 g/mol
Bewaar temperatuur: Room Temperature
MDL Nummer: MFCD27665386
CAS nummer: 760952-88-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR