2'3'-cGAMP ELISA Kit
Leverancier: Cayman Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
501700-96
CAYM501700-96EA 694 EUR
CAYM501700-96
2'3'-cGAMP ELISA Kit
Testen ELISAs
For the quantification of 2’3’-cGAMP in cell lysates.
  • Measure 2'3'-cGAMP in cell lysates
  • Assay 24 samples in triplicate or 36 samples in duplicate
  • Lower limit of detection (LLOD) is 9.6 pg/ml (0.01 pmol/ml)
  • Run overnight or incubate for just 2 hours without compromising sensitivity
  • Monitor the kinetics of 2'3'-cGAMP formation and hydrolysis in a biological setting
  • Identify compounds that modulate cGAS activity
Cayman’s 2’3’-cGAMP ELISA Kit is a competitive assay that can be used for quantification of 2’3’-cGAMP in cell lysates. This assay has a range from 100 ng/ml to 6,1 pg/ml with a midpoint of approximately 900 pg/ml (50% B/B0) and a sensitivity of approximately 85 pg/ml (80% B/B0). Powered by BIOLOG Life Science Institute.
Order Now
Specificaties
Environmentally Preferable
Target protein 2'3'-cGAMP
Primary antibody reactivity
Assaytype Competitive
Host
Formaat Pre-coated
Gevoeligheid 85,3 pg/ml
Detectiebereik 100 ng/ml - 6,1 pg/ml
Detectielimiet 9,6 pg/ml (0,01 pmol/ml)
Mid point 907,7 pg/ml