Bestatin hydrochloride ([S-(R*,S*)]-N-(3-Amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride)

Leverancier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

70520-10 70520-100 70520-5 70520-25
CAYM70520-10EA 140 EUR
CAYM70520-10 CAYM70520-100 CAYM70520-5 CAYM70520-25
Bestatin hydrochloride ([S-(R*,S*)]-N-(3-Amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride)
Bestatin hydrochloride ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine hydrochloride)
Formule: C₁₆H₂₅ClN₂O₄
MDL Nummer: MFCD00058004
CAS nummer: 65391-42-6

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR