Triphenylphosphine oxide

Leverancier: Cayman Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

9000289-25 9000289-50 9000289-500 9000289-1
CAYM9000289-25EA 25.2 EUR
CAYM9000289-25 CAYM9000289-50 CAYM9000289-500 CAYM9000289-1
Triphenylphosphine oxide
Triphenylphosphine oxide
Formule: C₁₈H₁₅OP
MW: 278.29 g/mol
Kookpunt: ∼360 °C (1013 hPa)
Smeltpunt: 155…158 °C
Dichtheid: 1.2 g/cm³ (20 °C)
Vlampunt: 180
MDL Nummer: MFCD00002080
CAS nummer: 791-28-6
EINECS: 212-338-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR