Drum racks
Leverancier: CEMO
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
7081 7639 7640 7642 10253 7734 7663 7664 7665 7670 7671 7672 7673 7674 7675 10254
CEMG7081EA 479 EUR
CEMG7081 CEMG7639 CEMG7640 CEMG7642 CEMG10253 CEMG7734 CEMG7663 CEMG7664 CEMG7665 CEMG7670 CEMG7671 CEMG7672 CEMG7673 CEMG7674 CEMG7675 CEMG10254
Drum racks
Opslag voor vaten
Drum racks for approved and space saving storage of barrels and other containers.
  • All drum racks are completely hotdip galvanised and have a corrosion free GRP sump pallet for long-term safe use
  • With general construction inspection approval no. Z-40.12-227 granted by DIBt-Berlin
  • Universally applicable for waste oil, lyes and acids
Order Now
Specificaties
Environmentally Preferable