Zoutzuur 36,5 - 38,0% NF, Multi-Compendial, meets analytical specification of ChP, BP, JP, Ph. Eur., J.T.Baker®

Leverancier: Avantor
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar


Management of Change (MOC) category = R

CH09-03EA 0 EUR
CH09-03 CH09-07
Zoutzuur 36,5 - 38,0% NF, Multi-Compendial, meets analytical specification of ChP, BP, JP, Ph. Eur., J.T.Baker®
Zoutzuur
Formule: HCl
Kookpunt: 110 °C (1013 hPa)
Smeltpunt: –30 °C
Dichtheid: 1,18 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00011324
CAS nummer: 7647-01-0
EINECS: 231-595-7
ADR: 8,II

Order Now

Specification Test Results

GMP Manufactured Product
Meets B.P. Chemical Specifications
Meets ChP Chemical Specifications
Meets E.P. Chemical Specifications
Meets J.P. Chemical Specifications
Meets N.F. Requirements
CAUTION: For Manufacturing, processing or repackaging
Bulk Pharmaceutical Chemical
NF - Identification Passes Test
NF - Residue on Ignition ≤ 0.008 %
NF - Bromide or Iodide Passes Test
NF - Free Bromine or Chlorine Passes Test
NF - Sulfate (SO₄) Passes Test
NF - Sulfite (SO₃) Passes Test
NF - Assay (HCl) 36.5 - 38.0 %
EP/BP - Identification A Passes Test
EP/BP - Identification B Passes Test
EP/BP - Identification C Passes Test
EP/BP - Appearance of Solution Passes Test
EP/BP - Free Chlorine (as Cl₂) ≤ 4 ppm
EP/BP - Sulfate (SO₄) ≤ 20 ppm
EP/BP - Residue on Evaporation ≤ 0.01 %
EP/BP - Assay (HCl) 35.0 - 39.0 %
JP - Assay (HCl) 35.0 - 38.0 %
JP - Identification Passes Test
JP - Residue on ignition, mg/10mL ≤ 1.0
JP - Bromide or Iodide Passes Test
JP - Bromine or Chlorine Passes Test
JP - Sulfate (SO₄) Passes Test
JP - Sulfite (SO₃) Passes Test
JP - Arsenic (As) ≤ 1 ppm
JP - Heavy Metals (as Pb) ≤ 5 ppm
JP - Mercury (Hg) ≤ 0.04 ppm
ChP - Assay 36.0 - 38.0 %
ChP - Identification Passes Test
ChP - Arsenic (As) ≤ 0.0001 %
ChP - Bromide or Iodide Passes Test
ChP - Free Chlorine or Bromine Passes Test
ChP - Heavy Metals ≤ 0.0002 %
ChP - Iron (Fe) ≤ 0.0001 %
ChP - Residue on Ignition ≤ 0.002 %
ChP - Sulfate ≤ 0.0005 %
ChP - Sulfite Passes Test
No Class 1, 2, 3 or other solvents are used or produced in the
manufacturing or purification of the product.
Elemental Impurities (USP 232, EP 5.20) - Information on elemental
impurities for this product is available on the associated Product
Regulatory Data Sheet and elemental impurity profile report.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR