D-Alanine

Leverancier: Chemicon
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

AB5777
CHMCAB5777EA 0 EUR
CHMCAB5777
D-Alanine
D-Alanine
Formule: C₃H₇NO₂
Kookpunt: 189 °C (1013 hPa)
Smeltpunt: 79 °C
MDL Nummer: MFCD00008077
CAS nummer: 338-69-2
EINECS: 206-418-1
Merck Index: 13,00203

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR