Tissue processing cassette, System II Hex
Leverancier: CELLPATH
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
EAD-0104-03A EAD-0105-03A EAD-0106-03A EAD-0107-03A EAD-0108-03A EAD-0109-03A
720-1892EA 75.3 EUR
720-1892 CLLPEAD-0105-03A 720-1893 720-1894 720-1895 720-1896
Tissue processing cassette, System II Hex
Histologie-cassettes Cassettes voor weefselbehandeling
The System II Hex Cassette differs from conventional round hole processing cassettes in that it has hexagonal instead of round perforations arranged in a honeycombed structure to better utilise the available surface area.
  • Available in different stock colours, which allow laboratories to colour code different types of workload or different applications
  • Manufactured from high–grade virgin acetyl resin
This unique arrangement increases the open area by approximately 13 percent thereby increasing reagent flow through and improving specimen staining andMore Product Information