,,1 component: Hg 0.5mg/l in HNO3 2% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 8 months.,, ,,Store under normal laboratory conditions, at temperatures between 15C to 25C,, 1 * 100 mL
Leverancier: CUSTOM MADE CHEMICALS LAB
,,1 component: Hg 0.5mg/l in HNO3 2% ,, ,,CRM, ISO 17034 and ISO 17025,, ,,Conservation 8 months.,, ,,Store under normal laboratory conditions, at temperatures between 15C to 25C,, 1 * 100 mL
New Product
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...
Meer About VWR
Bel voor prijs
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
Stock for this item is limited, but may be available in a warehouse close to you. Please make sure that you are logged in to the site so that available stock can be displayed. If the call is still displayed and you need assistance, please call us at 1-800-932-5000.
This product is marked as restricted and can only be purchased by approved Shipping Accounts. If you need further assistance, email VWR Regulatory Department at Regulatory_Affairs@vwr.com
-Additional Documentation May be needed to purchase this item. A VWR representative will contact you if needed.
Dit artikel is geblokkeerd voor uw organisatie. Neem contact op met uw inkoopafdeling voor meer informatie
Het orginele artikel is niet langer beschikbaar. Het alternatief dat wordt getoond is beschikbaar
Product(en) gemarkeerd met dit symbool worden niet meer verkocht cq verkocht tot het einde van de voorraad. Alternatieven zijn mogelijk beschikbaar door te zoeken met het VWR-catalogusnummer dat hierboven wordt vermeld. Voor meer informatie kunt u contact opnemen met customer service: 020-4808410.