Anti-HA tag Mouse Monoclonal Antibody (Biotin) [clone: 16B12]

Leverancier: Columbia Biosciences
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

B1-1722
COBSB1-1722EA 629 EUR
COBSB1-1722
Anti-HA tag Mouse Monoclonal Antibody (Biotin) [clone: 16B12]
Antilichamen
Type: Tag
Antigen: HA tag (HA tag)
Clonality: monoclonal
Clone: 16B12
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR