Anti-6X His tag Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: AD1.1.10]

Leverancier: Columbia Biosciences
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D3-1711-100 D3-1711-1MG
COBSD3-1711-100EA 629 EUR
COBSD3-1711-100 COBSD3-1711-1MG
Anti-6X His tag Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: AD1.1.10]
Antilichamen
Type: Tag
Antigen: 6X His tag (6X His tag)
Clonality: monoclonal
Clone: AD1.1.10
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR