Anti-IgG3 Goat Polyclonal Antibody (RPE (R-Phycoerythrin))

Leverancier: Columbia Biosciences
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D5-112-3
COBSD5-112-3EA 385 EUR
COBSD5-112-3
Anti-IgG3 Goat Polyclonal Antibody (RPE (R-Phycoerythrin))
Antilichamen
Type: Secondary
Antigen: IgG3 (Mouse) (mouse IgG3)
Clonality: polyclonal
Clone:
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR