Anti-IgG Goat Polyclonal Antibody (SureLight® P3)

Leverancier: Columbia Biosciences
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D7-114
COBSD7-114EA 623 EUR
COBSD7-114
Anti-IgG Goat Polyclonal Antibody (SureLight® P3)
Antilichamen
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: polyclonal
Clone:
Conjugation: SureLight® P3
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR