Anti-c-Myc tag Mouse Monoclonal Antibody (DyLight® 550) [clone: 9E10]

Leverancier: Columbia Biosciences
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D9-1714
COBSD9-1714EA 629 EUR
COBSD9-1714
Anti-c-Myc tag Mouse Monoclonal Antibody (DyLight® 550) [clone: 9E10]
Antilichamen
Type: Tag
Antigen: c-Myc tag (c-Myc tag)
Clonality: monoclonal
Clone: 90000000000
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR