Waterproof digital thermometer, DT400
Leverancier: COMARK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
DT400
COMADT400EA 37.9 EUR
COMADT400
Waterproof digital thermometer, DT400
Thermometers Digitale thermometers
Pocket digital thermometers used for checking food temperature and the final rinse cycle temperature of commercial dishwashers
  • Durable and field calibratable
  • Centigrade or Fahrenheit scales
  • BioCote Antimicrobial Protection
  • Sampling time of 1,5 seconds
  • Large digit display visible from the top of the instrument
Thin tip probe gives a response within 6 seconds. Maximum temperature hold button used for switching between the maximum reading and the current temperature reading.
One year warranty.
Certificaten: Complianced with NSF.