Pocket digital thermometer for dishwashers
Leverancier: COMARK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
KM14
COMAKM14EA 493 EUR
COMAKM14
Pocket digital thermometer for dishwashers
Thermometers Digitale thermometers
The KM14 dishwasher pocket thermometer incorporates a stainless steel penetration probe and can be placed inside a commercial dishwasher. A quick reading will show whether the dishwasher is operated at the correct temperature to sanitise the equipment.
  • Waterproof and field calibratable
  • Auto power off conserves battery life
  • BioCote® antimicrobial protection reduces the risk of cross-contamination
  • Combines accuracy with speed and durability
It is multi-purpose and can also be used as a conventional food penetration probe thermometer and is a topMore Product Information
Order Now
Specificaties
Environmentally Preferable