Digital clock/timer
Leverancier: COMARK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
UTL882
COMAUTL882EA 103 EUR
COMAUTL882
Digital clock/timer
Timers Digitale timers
This kitchen clock has a countdown capacity of 24 hours. All of its three functions − clock, timer and countdown − can operate simultaneously.
  • Built-in memory allows storage of a pre-set time
  • Count up in 1 second resolution up to 24 hours
Certificaten: Compliant with HACCP (hazard analysis and critical control points) procedures.
Leveringsinformatie: Incl. magnet, pocket clip, stand and battery.