Cleanroom wipes, pre-saturated, Polynit Heatseal, Prosat®
Leverancier: CONTEC
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PSPS0030
CONTPSPS0030EA 1030 EUR
CONTPSPS0030
Cleanroom wipes, pre-saturated, Polynit Heatseal, Prosat®
Schoonmaakdoekjes Cleanroom doekjes Vochtige cleanroom doekjes
These wipes are made of 100% knitted polyester pre-saturated with 100% semiconductor grade Isopropyl Alcohol (IPA).
  • No-run interlock knit for good solvent sorbency, abrasion resistance, and chemical resistance
  • Low in particles and extractables
  • Laundered and packaged in an ISO Class 4 cleanroom
Ideal for wiping exterior product surfaces and cleaning the interior surfaces of process tools or other equipment.
Colour: White.
Opgelet: *De product(en) op deze pagina vallen buiten het toepassingsgebied van de EU-richtlijn 2019/904