Measuring cylinders, low form, PP, with funnel top, Corning®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3022P-50 3022P-10 3022P-100 3022P-1L 3022P-250 3022P-2L 3022P-500 3022P-25
CORN3022P-50EA 195.95 EUR
CORN3022P-50 CORN3022P-10 CORN3022P-100 CORN3022P-1L CORN3022P-250 CORN3022P-2L CORN3022P-500 CORN3022P-25
Measuring cylinders, low form, PP, with funnel top, Corning®
Cilinders Maatcilinders
PP, doorschijnend, met schaalverdeling.
  • De cilinders zijn gemaakt met schaalverdeling in reliëf bij de belangrijkste maatstrepen
  • Met zeshoekige voet, gekalibreerd "TC, In"
  • Blootstelling aan temperaturen tot 60 °C kan de nauwkeurigheid niet beïnvloeden
Graduated cylinders Class B calibrated 'To Contain' (TC) according to DIN 12681/ISO 6706.
It is important to recognise the difference in specifications regarding DIN and ASTM standards. For reference, ASTM Class A and B specifications are more precise than DIN Class A and B specifications,More Product Information