Corning® closed system cell cubes
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3231 3232 3233
CORN3231EA 1476.38 EUR
CORN3231 CORN3232 CORN3233
Corning® closed system cell cubes
Microplaten Celcultuurplaten
Our new offering of CellCube 10, 25, 100 modules, circulation loops, and connectors allows for easy adherent cell scale-up that integrate seamlessly with AseptiQuik® and MPC connectors.
  • Standardized closed system designs allow flexible setup of any CellCube module and aseptic connection to an oxygenator (sold separately)
  • Sterile and ready-to-use offerings eliminate the time and complexity of assembling in-house
  • Industry standard AseptiQuik® S/G connectors allow for robust, single-use aseptic connections
The Corning CellCube system provides a fast,More Product Information