Micro-cavity plates, Corning® Elplasia®
Leverancier: Corning
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4440 4441 4442 4444 4445 4446 4447
CORN4440EA 593.82 EUR
CORN4440 CORN4441 391-1006 CORN4444 CORN4445 CORN4446 CORN4447
Micro-cavity plates, Corning® Elplasia®
Microplaten Celcultuurplaten
Elplasia plates enable researchers to generate high density of spheroids in a scaffold-free model. Use Corning Elplasia plates to generate, culture and analyse your spheroids all in a standard plate footprint.
  • Black opaque side walls reduce well-to-well cross-talk
  • Gamma irradiated
  • Suitable for fluorescent/luminescent assays
  • One year shelf life
These plates are compatible with many cell types and may be used across many applications including: Drug/high throughput screening, cancer/tumour biology, stem cell biology, cell therapy research and 3-D tissueMore Product Information
Order Now
Specificaties
Environmentally Preferable Lage impact op de fabricage
Milieu certificering RECs, ISO 14001 - Oneonta