Transport boxes, Securipack
Leverancier: E3 Cortex
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
FABRSECU1G1 FABRSECU03G1 FABRSECU05G1 FABRSECU2L5G1 FABRSECU5G1
CORTFABRSECU1G1EA 131 EUR
CORTFABRSECU1G1 CORTFABRSECU03G1 CORTFABRSECU05G1 CORTFABRSECU2L5G1 CORTFABRSECU5G1
Transport boxes, Securipack
Dozen Transportdozen
This Securipack packages are ready for use in the transport of samples of hazardous materials, except for Classes 7 and 6.2.
  • Very simple to use, this package eliminates risk posed by error or oversight
  • Mechanical high strength, in particular in the stacking by the juxtaposition of double walls on all the faces
  • High absorption power of the shocks on all the faces, guaranteeing a perfect protection in case of an accidental fall
Universal application for products regulated except for the radioactive and infectious products.
Protects against theMore Product Information