Sleeves for ground joints, rigid
Leverancier: COWIE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
020.134 020.145 020.110 020.124 020.129 020.155 020.119.1 020.114.1
COWI020.134EA 14.1 EUR
COWI020.134 COWI020.145 COWI020.110 COWI020.124 COWI020.129 COWI020.155 COWI020.119.1 COWI020.114.1
Sleeves for ground joints, rigid
Geslepen Geslepen hulzen
Made of PTFE, grease-free, high vacuum full length rigid sleeves for standard glass joints, with substantial shoulder for ease of handling and repeated use.
  • Rigid sleeves and adapters are inert
  • Usable to 280˚C high vacuum
  • The leakage rate is less
  • High vacuum performance is obtained by transverse sealing rings