CPC® NS6 Quick Fittings, ³/₈" Hose Barb Valved In-Line Couplings
Leverancier: CPC COLDER
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
NS6D17006 NS6D22006 NS6D42006
CPCCNS6D17006EA 259 EUR
CPCCNS6D17006 CPCCNS6D22006 CPCCNS6D42006
CPC® NS6 Quick Fittings, ³/₈" Hose Barb Valved In-Line Couplings
Fittingen en connectoren
NS6 Series couplings feature non-spill valves at a great price. Use the NS6 when even a few drops of spillage pose problems regarding safety, media cost or environmental regulations. These innovative couplings are lightweight, chemically resistant and easy to use.
  • Non-spill design - Disconnect under pressure with no spills
  • Colour coding - Instant visual differentiation of media lines
  • Glass-filled polypropylene - Durable and compatible with many chemicals
  • CPC thumb latch - One-hand connection and disconnection
The non-spill design virtually eliminatesMore Product Information