Screw gate carabiner, Steel, Protecta®, 3M™
Leverancier: CAPITAL SAFETY
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
AJ501
CPSDAJ501EA 0 EUR
CPSDAJ501
Screw gate carabiner, Steel, Protecta®, 3M™
Valbescherming Karabijnhaken
The Protecta® Screw gate carabiner is ideal in working situations where opening is occasional during the working day, as the gate can be opened by pushing it onto the anchorage point. The gate can then be manually locked once the carabiner holds the weight of the device.
  • Manual locking design is ideal for occasional opening during the working week
  • For use in all working at height applications
  • Durable and lightweight
  • Corrosion-resistant
Anchorage connectors are a critical component in a fall protection system, as they securely attach the user's systemMore Product Information