Supelco® TraceCERT® Organic Reference Standard, Acequinocyl
Leverancier: Merck
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
CRM52422-50MG
CRM52422-50MGEA 138 EUR
CRM52422-50MG
Supelco® TraceCERT® Organic Reference Standard, Acequinocyl
Standaarden Organische standaarden
Certificaten: This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI e.g. NIST or NMIJ. Certified content by quantitative NMR including uncertainty and expiry date are given on the certificate.
Opgelet: P260 - P264 - P273 - P280 - P302 + P352 - P308 + P311