E.Z.N.A.® Plant DNA DS kit
Leverancier: OMEGA BIO-TEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
D2411-00 D2411-01
D2411-00EA 25 EUR
D2411-00 D2411-01
E.Z.N.A.® Plant DNA DS kit
Reagentia voor nucleïnezuren Nucleic Acid Purification Kits and Reagents
The E.Z.N.A.® Plant DNA DS Mini Kit is designed for efficient recovery of genomic DNA up to 30 kb in size from fresh, frozen, or dried plant tissue samples rich in polysaccharides, polyphenols or having a lower DNA content. Up to 50 mg wet tissue can be processed in less than 1 hour. The system combines the reversible nucleic acid-binding properties of the HiBind® matrix with the speed and versatility of spin column technology to eliminate polysaccharides, phenolic compounds, and enzyme inhibitors from plant tissue lysates. Purified DNA is suitable for PCR, restriction digestion, and hybridisation applications.
  • Rapid - homogeniser columns allow for faster processing
  • Versatile - reliable results from a variety of sample types
  • Safe - no organic extractions
  • High quality - purified DNA suitable for most applications
This procedure relies on the well-established properties of the cationic detergent, cetyltrimethyl ammonium bromide (CTAB), in conjunction with the unique binding system to increase yields and provide high-quality DNA. The system eliminates the need for chloroform extractions traditionally associated with with CTAB-based lysis methods. Samples are homogenised and lysed in a high salt buffer containing CTAB, binding conditions are adjusted, and DNA is purified using a E-Z 96® DNA plate. Salts, proteins, and other contaminants are removed to yield high-quality genomic DNA suitable for downstream applications such as endonuclease digestion, thermal cycle amplification, and hybridisation applications.