E.Z.N.A.® Plasmid Mini kit I

Leverancier: OMEGA BIO-TEK
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

D6943-02 D6943-01 D6942-00 D6943-00 D6942-01 D6942-02
D6943-02EA 237 EUR
D6943-02 D6943-01 D6942-00 D6943-00 D6942-01 D6942-02
E.Z.N.A.® Plasmid Mini kit I
Reagentia voor nucleïnezuren Nucleic Acid Purification Kits and Reagents

The E.Z.N.A.® Plasmid Mini Kit I is designed to isolate up to 30 µg of high-quality plasmid DNA from 1 to 5 ml bacterial cultures in 30 minutes or less. Plasmid DNA purification is simplified with HiBind® Mini Column technology into three quick steps: Bind, wash, and elute. Purified plasmid DNA is immediately ready for a wide variety of downstream applications such as routine screening, restriction enzyme digestion, and manual/automated fluorescent DNA sequencing. E.Z.N.A.® Plasmid Mini Kit I is available in either V-spin or Q-spin column format. V-spin column formats have an attached lid while Q-spin columns do not. Both versions can be used for centrifugation or vacuum protocols.


  • Purification of plasmid DNA in 30 minutes or less
  • No phenol/chloroform extractions
  • DNA suitable for downstream applications

Order Now

Specificaties

Omschrijving E.Z.N.A.® Plasmid Mini Kit I (V-Spin column) E.Z.N.A.® Plasmid Mini Kit I (Q-Spin column)
Environmentally Preferable
Applicatie
Target molecule Plasmid Plasmid
Refine target molecule Plasmid Plasmid
Sample type Bacteria Bacteria
Method / Format Column (silica-based) Column (silica-based)
Prep size Mini Mini
Monstergrootte 1 - 5 ml 1 - 5 ml

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR